Learn More
Velp Scientifica™ Glass Beakers
Product Code. 15359624 Shop All Velp Products
Click to view available options
Capacity (Metric):
1000 mL
Unit Size:
Each
Product Code. 15359624
Brand: Velp Scientifica™ A00001001
Only null left
Add to basket
Add to basket
Excellent for holding solutions during the flocculation process. Velp Scientifica™ Glass Beakers are for use with FC S and JLT Series Flocculators.
  • Excellent chemical resistance
  • Easy to use
  • Clear glass
  • Graduation marks
Specifications
Capacity (Metric) 1000 mL
For Use With (Equipment) FC S and JLT Series Flocculators
Type Beaker