missing translation for 'onlineSavingsMsg'
Learn More

Hanna Instruments™ Glass BNC Connector Combination pH Electrode

Product Code. 10172701
Change view
Click to view available options
pH Range:
0 to 14
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. pH Range Unit Size
10172701 0 to 14 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10172701 Supplier Hanna Instruments™ Supplier No. HI1043B
  • ONLINE EXCLUSIVE PRICE
    498.74 €
    276.64 € / Pack of 1
    Save 222.10 €
    44% Off
Estimated Shipment: 13-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

Connector Types BNC
Type BNC Connector Combination pH Electrode
For Use With (Application) Used at elevated temperatures the resistance decreases to approach the that of GP glass
Material Glass
pH Range 0 to 14
Temperature (Metric) Range 0°C to 100°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.