Learn More
Hirschmann™ Class A Volumetric Flasks with Poly Stopper
Achieve precise standard solutions. Hirschmann™ Class A Volumetric Flasks with Poly Stopper is standardized to contain or to deliver a definite amount of liquid. Flask has silver diffusion ink with brown graduations.
40.10 € - 337.00 €
Specifications
Certifications/Compliance | DIN EN ISO 1042 |
---|---|
Closure Material | Polyethylene |
Class | A |
Product Code | Brand | Capacity (Metric) | Standard Taper Joint | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | Standard Taper Joint | Price | Quantity & Availability | ||||
10030052
|
Hirschmann™
2822192 |
1000 mL | 19/32 NS Standard Ground |
65.90 €
Each |
Estimated Shipment: 14-06-2024
Log in to see stock. |
||||
13197949
|
Hirschmann™
2822152 |
5 mL | 7/16 NS Standard Ground |
40.10 €
Pack of 2 |
Estimated Shipment: 04-06-2024
Log in to see stock. |
||||
12628675
|
Hirschmann™
2822180 |
100 mL | 12/21 NS Standard Ground |
50.30 €
Pack of 2 |
Estimated Shipment: 31-05-2024
Log in to see stock. |
||||
11952043
|
Hirschmann™
2822181 |
100 mL | 14/23 NS Standard Ground |
52.53 €
Pack of 2 |
Estimated Shipment: 04-06-2024
Log in to see stock. |
||||
11912043
|
Hirschmann™
2822161 |
10 mL | 10/19 NS Standard Ground |
40.10 €
Pack of 2 |
Estimated Shipment: 04-06-2024
Log in to see stock. |
||||
11962043
|
Hirschmann™
2822185 |
200 mL | 14/23 NS Standard Ground |
57.10 €
Pack of 2 |
Estimated Shipment: 04-06-2024
Log in to see stock. |
||||
11922043
|
Hirschmann™
2822165 |
20 mL | 10/19 NS Standard Ground |
41.70 €
Pack of 2 |
Estimated Shipment: 04-06-2024
Log in to see stock. |
||||
10174924
|
Hirschmann™
2822178 |
50 mL | NS 14/23 |
46.20 €
Pack of 2 |
Estimated Shipment: 04-06-2024
Log in to see stock. |
||||
10758811
|
Hirschmann™
2822194 |
2000 mL | 29/32 NS Standard Ground |
82.90 €
Each |
Estimated Shipment: 27-06-2024
Log in to see stock. |
||||
10245522
|
Hirschmann™
2822166 |
20 mL | 12/21 NS Standard Ground |
41.80 €
Pack of 2 |
Estimated Shipment: 04-06-2024
Log in to see stock. |
||||
11932043
|
Hirschmann™
2822170 |
25 mL | 10/19 NS Standard Ground |
41.70 €
Pack of 2 |
Estimated Shipment: 31-05-2024
Log in to see stock. |
||||
10386342
|
Hirschmann™
2822171 |
25 mL | 12/21 NS Standard Ground |
45.90 €
Pack of 2 |
Estimated Shipment: 04-06-2024
Log in to see stock. |
||||
11982043
|
Hirschmann™
2822190 |
500 mL | 19/26 NS Standard Ground |
70.20 €
Pack of 2 |
Estimated Shipment: 04-06-2024
Log in to see stock. |
||||
11902043
|
Hirschmann™
2822153 |
5 mL | 10/19 NS Standard Ground |
40.10 €
Pack of 2 |
Estimated Shipment: 27-06-2024
Log in to see stock. |
||||
11942043
|
Hirschmann™
2822175 |
50 mL | 12/21 NS Standard Ground |
45.50 €
Pack of 2 |
Estimated Shipment: 04-06-2024
Log in to see stock. |
||||
11972043
|
Hirschmann™
2822186 |
250 mL | 14/23 NS Standard Ground |
59.10 €
Pack of 2 |
Estimated Shipment: 04-06-2024
Log in to see stock. |
||||
10315231
|
Hirschmann™
2822193 |
1000 mL | 24/29 NS Standard Ground |
60.70 €
Each |
Estimated Shipment: 04-06-2024
Log in to see stock. |
||||
10151742
|
Hirschmann™
2822197 |
5000 mL | 34/35 NS Standard Ground |
181.00 €
Each |
Estimated Shipment: 13-06-2024
Log in to see stock. |
||||
10611923
|
Hirschmann™
2822198 |
10,000 mL | 45/40 NS Standard Ground |
337.00 €
Each |
Estimated Shipment: 18-07-2024
Log in to see stock. |
||||
Description
- With polyethylene stopper
- With amber stain graduation
- Acc. to DIN EN ISO 1042
- Corrosion-free and resistant to thermal shock
- Conformity certified, with dated batch identification
Specifications
DIN EN ISO 1042 | |
Polyethylene | |
A |