Learn More
Hirschmann™ Class A Volumetric Flasks with Poly Stopper
Achieve precise standard solutions. Hirschmann™ Class A Volumetric Flasks with Poly Stopper is standardized to contain or to deliver a definite amount of liquid. Flask has silver diffusion ink with brown graduations.
Brand:  Hirschmann™ 2822181
Additional Details : Weight : 0.25000kg
 View more versions of this product
Product Code. 11952043
  • 52.53 € / Pack of 2
Estimated Shipment: 24-05-2024
to see stock.
Add to basket
Description
Description
  • With polyethylene stopper
  • With amber stain graduation
  • Acc. to DIN EN ISO 1042
  • Corrosion-free and resistant to thermal shock
  • Conformity certified, with dated batch identification
Specifications
Specifications
DIN EN ISO 1042
100 mL
170 mm
14/23 NS Standard Ground
Yes
Polyethylene
Borosilicate Glass, Duran Glass
±0.1 mL
±0.1 mL
A