missing translation for 'onlineSavingsMsg'
Learn More

Witeg™ Reagent Bottles Amber Glass Wide Mouth

Product Code. 15274638
Change view
Click to view available options
Capacity (Metric):
1000 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric)
15274638 1000 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15274638 Supplier Witeg™ Supplier No. 5 895 001
  • ONLINE EXCLUSIVE PRICE
    121.66 €
    93.85 € / Pack of 12
    Save 27.81 €
    22% Off
 Restricted Item
In Stock
Add to basket
Edge
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Soda glass reagent bottles have a wide neck for easy filling and retrieving samples, they are supplied with a black PP screw cap.

  • Amber glass
  • Wide neck
  • PP screw cap included

Specifications

Capacity (Metric) 1000 mL
Color Amber
Material Glass
Quantity 12 Pk.
Closure Color Black
Closure Included Yes
Closure Type Screw Cap
Shape Round
Product Type Wide Mouth Bottle
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.