Learn More
Hirschmann™ ringcaps™ Disposable Capillary Pipettes
Product Code. 11722313
Change view
Click to view available options
Volume (Metric):
1 to 5 μL
10 μL
100 μL
100 to 200 μL
20 μL
20 to 40 μL
44.7 μL
50 μL
50 to 100 μL
Includes:
1 Ring Mark
2 Ring Marks
5 Ring Marks
Unit Size:
Pack of 1000
Pack of 400
10 product options available for selection
Product selection table with 10 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) Includes unitSize
11722313 20 μL 1 Ring Mark Pack of 1000
11762293 1 to 5 μL 5 Ring Marks Pack of 1000
11762313 50 to 100 μL 2 Ring Marks Pack of 1000
11772313 100 μL 1 Ring Mark Pack of 1000
15559035 44.7 μL 1 Ring Mark Pack of 1000
11772323 100 to 200 μL 2 Ring Marks Pack of 400
11712313 10 μL 2 Ring Marks Pack of 1000
11702313 10 μL 1 Ring Mark Pack of 1000
11742313 20 to 40 μL 2 Ring Marks Pack of 1000
11752313 50 μL 1 Ring Mark Pack of 1000
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 10 options available.
10 options
Product Code. 11722313 Supplier Hirschmann™ Supplier No. 9600120
  • ONLINE EXCLUSIVE PRICE
    129.15 €
    114.20 € / Pack of 1000
    Save 14.95 €
    11% Off
 Restricted Item
Estimated Shipment: 27-10-2026
Add to basket
Add to basket
Excellent for working with small samples in the laboratory. Hirschmann Instruments™ ringcaps™ Disposable Capillary Pipettes can be utilized for a wide range of scientific and medical applications.
  • With ringmark
  • Conformity certified
  • Manufactured from durable DURAN™ glass
  • Acc. to ISO 7550, CE acc. IVD 98/79 EC
Specifications
Accuracy 0.25%
Certifications/Compliance ISO 7550, CE, IVD 98/79 EC
Material DURAN
Graduations 20 μL
Includes 1 Ring Mark
Volume (Metric) 20 μL
Type Capillary Pipette
Disposable Single-use