Learn More
Hirschmann™ ringcaps™ Disposable Capillary Pipettes
Product Code. 11732313
Change view
Click to view available options
Volume (Metric):
25 μL
Unit Size:
Pack of 1000
Product Code. Volume (Metric) Unit Size
11732313 25 μL Pack of 1000
1 options
Product Code. 11732313 Supplier Hirschmann™ Supplier No. 9600125
  • ONLINE EXCLUSIVE PRICE
    136.00 €
    114.20 € / Pack of 1000
    Save 21.80 €
    16% Off
 Restricted Item
In Stock
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Excellent for working with small samples in the laboratory. Hirschmann Instruments™ ringcaps™ Disposable Capillary Pipettes can be utilized for a wide range of scientific and medical applications.
  • With ringmark
  • Conformity certified
  • Manufactured from durable DURAN™ glass
  • Acc. to ISO 7550, CE acc. IVD 98/79 EC
Specifications
Accuracy 0.25%
Certifications/Compliance ISO 7550, CE, IVD 98/79 EC
Material Glass
Includes Ring Mark
Volume (Metric) 25 μL
Type Capillary Pipette
Disposable Single-use