Learn More
Eppendorf™ Glass Cuvette
Product Code. 15204576 Shop All Eppendorf Products
Change view
Click to view available options
Description:
Model: 4011, semi-micro
Model: 4052, flow-through
Unit Size:
Each
Product Code. Description unitSize
15204576 Model: 4052, flow-through Each
15284566 Model: 4011, semi-micro Each
2 options
Product Code. 15204576
Brand: Eppendorf™ 4052102001
Only null left
Add to basket
Add to basket
Designed for a variety of applications
  • Optical glass composition
  • Available in a variety of models and sizes
TRUSTED_SUSTAINABILITY
Specifications
Type Cuvette
Description Model: 4052, flow-through
Material Glass
Quantity 1 Each
For Use With (Equipment) Spectrophotometer
Width (Metric) 10 mm
For Use With (Application) Spectroscopy