missing translation for 'onlineSavingsMsg'
Learn More

Cole-Parmer™ Glass Drying Tray

Product Code. 15322812 Shop All Cole-Parmer Products
Click to view available options
Dimensions (L x W x H):
200 x 200 x 50 mm
Unit Size:
Pack of 4
This item is not returnable. View return policy

Product Code. 15322812

Brand: Cole-Parmer™ 3458822

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Heavy duty trays endure hot air or steam sterilisation

  • Pyrex 3170 heavy duty trays
  • Ideal for drying, staining or developing and are autoclavable
  • Can be used with electrophoresis gels and paper chromatography
  • Do not place over direct heat

Specifications

Capacity (Metric) 2 L
Dimensions (L x W x H) 200 x 200 x 50 mm
Color Clear
Material Glass
Type Glass tray
For Use With (Application) Drying, staining or developing, electrophoresis gels and paper chromatography (Do not place over direct heat)
Autoclavable Autoclavable
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.