missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Glass-Encased Stir Bar

Product Code. 11547792 Shop All Fisherbrand Traceable Products
Change view
Click to view available options
Length (Metric):
6 mm
12 mm
19 mm
25 mm
45 mm
60 mm
Diameter (Metric):
5 mm
6 mm
8 mm
Unit Size:
Pack of 5
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Length (Metric) Diameter (Metric) Unit Size
11547792 12 mm 5 mm Pack of 5
11537792 6 mm 5 mm Pack of 5
11567792 25 mm 6 mm Pack of 5
11848902 45 mm 8 mm Pack of 5
11858902 60 mm 8 mm Pack of 5
11557792 19 mm 5 mm Pack of 5
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Product Code. 11547792
In Stock
Add to basket
Edge
Add to basket

missing translation for 'validMainframeMsg'

Use with very abrasive media that may erode PTFE. The glass-encased magnetic stir bar is cylindrical for a wide variety of uses.

  • Stable, general-purpose magnetic stir bar
  • Use with very abrasive media that may erode PTFE
  • Autoclavable

Specifications

Autoclavable Autoclavable
Certifications/Compliance Certificate of Conformance, Certificate of Analysis, Product Data Sheet, Material Safety Data Sheet (MSDS)
Color Clear
Description Glass encapsulated Alnico V magnet core
For Use With (Equipment) Containers & Vessels / Stirrer Hot Plates
Diameter (Metric) 5 mm
Length (Metric) 12 mm
Quantity 5/Pk.
Shape Cylinder
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.