Learn More
Merck Glass Fiber Filters Without Binder
Choose from six grades of filters. Glass Fiber Filters without Binders are made of borosilicate microfiber glass. Available in multiple pore sizes and flow rates for a variety of filtration and analysis applications utilizing depth filters. Autoclavable.
Brand: Merck AP4002500
Code : NEW
Additional Details : Weight : 0.10000kg
Description
For prefiltration and contamination analysis
Millipore fibrous disc filters are available in a wide range of flow rates and throughput capacities. Our Glass Fiber filters are available with or without binder resins and can be sterilized by EO, gamma, or autoclave (121 °C at 1 bar).Glass Fiber Filters without Binder Resin:
The filters without binder resin retain their structural integrity without weight loss when heated up to 500 °C and can therefore be used in gravimetric analysis as well as for the filtration of hot gases.
Type APFF
•Use for filtering extremely fine precipitates such as protein, nucleic acids, or serum precipitates
•Recommended for USEPA method 1311 for TCLP analysis
Type AP40
•Recommended for Standard Methods Total Suspended Solids 2540D
•Recommended for USEPA method 1311 for TCLP analysis
•Maintains structural integrity without weight loss when ignited to 550 °C (1022 °F) after sample filtration
•Recommended for determining volatile suspended matter in wastewater and industrial effluents
Specifications
44.5 L/min/ cm2 | |
Autoclavable | |
Depth Filter | |
0.7 μm | |
6 mL/min/ cm2 | |
AP40 | |
Plain | |
550°C | |
475 μm | |
Hydrophilic | |
2.5 cm |
50 mm of H2O | |
White | |
Borosilicate Glass Microfiber | |
Sterile | |
Used for contamination analysis | |
Hydrophilic | |
1022°F | |
0.018 in. | |
Filter Membranes | |
0.98 in. | |
2.5 cm |