Learn More
Merck Glass Fiber Filters Without Binder
Choose from six grades of filters. Glass Fiber Filters without Binders are made of borosilicate microfiber glass. Available in multiple pore sizes and flow rates for a variety of filtration and analysis applications utilizing depth filters. Autoclavable.
58.95 € - 735.00 €
Specifications
Color | White |
---|---|
Filter Type | Depth Filter |
Pore Size | 0.7 μm |
Surface Type | Plain |
Type | Filter Membranes |
Product Code | Brand | Diameter (Metric) | Grade | Flow Rate | For Use With (Application) | Price | Quantity & Availability | ||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Diameter (Metric) | Grade | Flow Rate | For Use With (Application) | Price | Quantity & Availability | ||
10635591
|
Merck
AP4003705 |
3.7 cm | AP40 | 6 mL/min/ cm2 | Determination of volatile suspended matter in wastewater and industrial effluents |
296.00 €
Pack of 500 |
Estimated Shipment: 06-06-2024
Log in to see stock. |
||
10598442
|
Merck
AP4002405 |
2.4 cm | AP40 | 6 mL/min/ cm2 | Determination of volatile suspended matter in wastewater and industrial effluents |
245.00 €
Pack of 500 |
Estimated Shipment: 06-06-2024
Log in to see stock. |
||
10379980
|
Merck
AP4014250 |
14.2 cm | AP40 | 6 mL/min/ cm2 | Determination of volatile suspended matter in wastewater and industrial effluents |
241.00 €
Pack of 50 |
Estimated Shipment: 06-06-2024
Log in to see stock. |
||
10498603
|
Merck
AP4002500 |
2.5 cm | AP40 | 6 mL/min/ cm2 | Used for contamination analysis |
58.95 €
Pack of 100 |
Estimated Shipment: 06-06-2024
Log in to see stock. |
||
10004570
|
Merck
AP4004705 |
4.7 cm | AP40 | 6 mL/min/ cm2 | Determining volatile suspended matter in wastewater and industrial effluents |
335.00 €
Pack of 500 |
Estimated Shipment: 20-06-2024
Log in to see stock. |
||
10572844
|
Merck
AP4004700 |
4.7 cm | AP40 | 6 mL/min/ cm2 | Determination of volatile suspended matter in wastewater and industrial effluents |
91.15 €
Pack of 100 |
Estimated Shipment: 30-05-2024
Log in to see stock. |
||
10588442
|
Merck
AP4001000 |
1 cm | AP40 | 6 mL/min/ cm2 | Determination of volatile suspended matter in wastewater and industrial effluents |
65.20 €
Pack of 100 |
Estimated Shipment: 06-06-2024
Log in to see stock. |
||
10437221
|
Merck
APFF14250 |
14.2 cm | APFF | 1.4 mL/min/ cm2 | Use for filtering extremely fine precipitates such as protein, Nucleic Acids, or Serum Precipitates |
600.00 €
Pack of 50 |
Estimated Shipment: 06-06-2024
Log in to see stock. |
||
10697121
|
Merck
APFF02500 |
2.5 cm | APFF | 1.4 mL/min/ cm2 | Use for filtering extremely fine precipitates such as protein, Nucleic Acids, or Serum Precipitates |
175.00 €
Pack of 100 |
Estimated Shipment: 06-06-2024
Log in to see stock. |
||
11758792
|
Merck
APFF12450 |
124 mm | APFF | 1.4 mL/min/ cm2 (water) | Fine particulate and microorganism removal than APFA filters |
431.00 €
Pack of 50 |
Estimated Shipment: 01-07-2024
Log in to see stock. |
||
10480021
|
Merck
APFF09050 |
9 cm | APFF | 1.4 mL/min/ cm2 | Use for filtering extremely fine precipitates such as protein, Nucleic Acids, or Serum Precipitates |
212.00 €
Pack of 50 |
Estimated Shipment: 06-06-2024
Log in to see stock. |
||
10340131
|
Merck
AP4009000 |
9 cm | AP40 | 6 mL/min/ cm2 | Determination of volatile suspended matter in wastewater and industrial effluents |
166.00 €
Pack of 100 |
Estimated Shipment: 06-06-2024
Log in to see stock. |
||
10498793
|
Merck
AP4007000 |
7 cm | AP40 | 6 mL/min/ cm2 | Used for contamination analysis |
130.00 €
Pack of 100 |
Estimated Shipment: 06-06-2024
Log in to see stock. |
||
10257770
|
Merck
APFF04700 |
4.7 cm | APFF | 1.4 mL/min/ cm2 | Use for filtering extremely fine precipitates such as protein, Nucleic Acids, or Serum Precipitates |
185.00 €
Pack of 100 |
Estimated Shipment: 06-06-2024
Log in to see stock. |
||
11768772
|
Merck
AP408X105 |
20.3 cm | AP40 | 6 mL/min/ cm2 | Determining volatile suspended matter in wastewater and industrial effluents |
735.00 €
Pack of 50 |
Estimated Shipment: 06-06-2024
Log in to see stock. |
||
Description
For prefiltration and contamination analysis
OverviewMillipore fibrous disc filters are available in a wide range of flow rates and throughput capacities. Our Glass Fiber filters are available with or without binder resins and can be sterilized by EO, gamma, or autoclave (121 °C at 1 bar).Glass Fiber Filters with Binder Resin:Millipore filters with binder resin have superior wet strength and are excellent for qualitative analysis and prefiltration, especially for heavily contaminated liquids. They are also widely used for clarification of aqueous solutions.Type AP15•Lowest dirt-holding capacity •Recommended prefilter for 0.2 to 0.6 μm filters Type AP20•Lower retention, higher dirt-holding capacity than AP15 filters •Recommended prefilter for 0.8 to 8.0 μm filters •Use upstream to protect type AP15 filters Type AP25•Added thickness yields same retention and greater dirt-holding capacity as AP20 •Recommended prefilter for 0.9 to 8 μm filters, especially for proteinaceous and heavily contaminated liquids •Use upstream to protect type AP15 filters Glass Fiber Filters without Binder Resin:The filters without binder resin retain their structural integrity without weight loss when heated up to 500 °C and can therefore be used in gravimetric analysis as well as for the filtration of hot gases.Type APFA•Retains fine particles with good efficiency, even at high flow rates •Recommended for monitoring wastewater and collecting suspended particles in gases (e.g., fumes from chimneys) •Use in biochemical applications (e.g., collection of cells and filtration of protein or nucleic acid precipitates) Type APFB•Higher mechanical strength when wet and greater loading capacity than type APFA filters •Use for liquid clarification, quantification of solids in suspensions of fine particles, and scintillation counting Type APFC•Greater retention, especially for fine particulate and microorganism removal than type APFA filters •Use for determining total suspended solids in drinking water •Use for filtering proteins or nucleic acid TCA precipitates and for collecting cells and microorganisms Type APFD•Thick filter with high flow and low retention •Use for clarifying suspensions containing particulates greater than 1.0 μm Type APFF•Use for filtering extremely fine precipitates such as protein, nucleic acids, or serum precipitates •Recommended for USEPA method 1311 for TCLP analysis Type AP40•Recommended for Standard Methods Total Suspended Solids 2540D •Recommended for USEPA method 1311 for TCLP analysis •Maintains structural integrity without weight loss when ignited to 550 °C (1022 °F) after sample filtration •Recommended for determining volatile suspended matter in wastewater and industrial effluentsSpecifications
White | |
0.7 μm | |
Filter Membranes |
Depth Filter | |
Plain |