missing translation for 'onlineSavingsMsg'
Learn More

Merck Glass Fiber Filters Without Binder

Choose from six grades of filters. Glass Fiber Filters without Binders are made of borosilicate microfiber glass. Available in multiple pore sizes and flow rates for a variety of filtration and analysis applications utilizing depth filters. Autoclavable.

Brand:  Merck AP408X105

Code : NEW

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 11768772

  • 735.00 € / Pack of 50
Estimated Shipment: 06-06-2024
to see stock.
Add to basket

Description

Description

For prefiltration and contamination analysis

Millipore fibrous disc filters are available in a wide range of flow rates and throughput capacities. Our Glass Fiber filters are available with or without binder resins and can be sterilized by EO, gamma, or autoclave (121 °C at 1 bar).
Glass Fiber Filters without Binder Resin:
The filters without binder resin retain their structural integrity without weight loss when heated up to 500 °C and can therefore be used in gravimetric analysis as well as for the filtration of hot gases.

Type APFF
•Use for filtering extremely fine precipitates such as protein, nucleic acids, or serum precipitates
•Recommended for USEPA method 1311 for TCLP analysis

Type AP40
•Recommended for Standard Methods Total Suspended Solids 2540D
•Recommended for USEPA method 1311 for TCLP analysis
•Maintains structural integrity without weight loss when ignited to 550 °C (1022 °F) after sample filtration
•Recommended for determining volatile suspended matter in wastewater and industrial effluents

Specifications

Specifications

44.5 L/min/ cm2
Autoclavable
Depth Filter
0.7 μm
Sterile
Determining volatile suspended matter in wastewater and industrial effluents
Hydrophilic
1022°F
0.018 in.
Filter Membranes
8 in.
20.3 cm
50 mm of H2O
White
Borosilicate Glass Microfiber
0.7 μm
6 mL/min/ cm2
AP40
Plain
550°C
475 μm
Hydrophilic
20.3 cm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers