Learn More
Brady™ Glass-Filled Nylon Miniature Circuit Breaker Lockouts
Product Code. 11771945
Click to view available options
Color:
Red
Red/Black
Unit Size:
1 pack
Each
Product Code. 11771945
Brand: Brady™ 090850
Only null left
Add to basket
Add to basket
A safe and effective method for locking out European Circuit Breakers.
  • Requires no tools to install
  • Lockout easily installs with the aid of a push button
  • Tie-bar style lockout has a thumb wheel for quick application
Specifications
Product Type Miniature Circuit Breaker Lockouts
Alternate Part Number ELEC. CIRCUIT BREAKER L/O EURO POW EA
Color Red/Black
Material Glass-filled Nylon
Certifications/Compliance UL 94 HB
Description Miniature Circuit Breaker Lockouts - Pin-Out Wide
Diameter (Metric) Shackle 7 mm
Material (Shackle) Glass-filled Nylon
Quantity 1 Ea.