Learn More
Hirschmann™ Glass Inserts
Product Code. 30158520
Change view
Click to view available options
No. per Pack:
384/Pk.
Unit Size:
Pack of 19200
Product Code. No. per Pack Unit Size
30158520 384/Pk. Pack of 19200
1 options
Product Code. 30158520 Supplier Hirschmann™ Supplier No. 9240584
  • ONLINE EXCLUSIVE PRICE
    9090.00 €
    5600.00 € / Pack of 19200
    Save 3490.00 €
    38% Off
Estimated Shipment: 23-11-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Glass inserts for Hirschmann™ plates formats. Glass providing excellent chemical inertness, no leaching of plastics.
  • Loose glass inserts
  • Highly pure
  • Chemically inert
Specifications
Material DURAN™ Glass
No. per Pack 384/Pk.
For Use With (Equipment) 384-well Hirschmann plates
Product Type Glass Inserts