Learn More
Hirschmann™ minicaps™ Micro Capillary Pipettes, Disposable
Facilitates error-free capillary filling in the laboratory
105.00 € - 113.00 €
Product Code | Brand | Volume (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Volume (Metric) | Price | Quantity & Availability | |||||
11871060
|
Hirschmann™
9000100 |
0.5 μL |
106.00 €
Pack of 1000 |
Estimated Shipment: 28-05-2024
Log in to see stock. |
|||||
11780055
|
Hirschmann™
9000199 |
100 μL |
113.00 €
Pack of 1000 |
Estimated Shipment: 11-07-2024
Log in to see stock. |
|||||
11871050
|
Hirschmann™
9000110 |
10 μL |
106.00 €
Pack of 1000 |
Estimated Shipment: 06-06-2024
Log in to see stock. |
|||||
11821050
|
Hirschmann™
9000101 |
1 μL |
106.00 €
Pack of 1000 |
In Stock
Log in to see stock. |
|||||
11722293
|
Hirschmann™
9000120 |
20 μL |
106.00 €
Pack of 1000 |
Estimated Shipment: 28-05-2024
Log in to see stock. |
|||||
11881060
|
Hirschmann™
9000125 |
25 μL |
113.00 €
Pack of 1000 |
Estimated Shipment: 11-07-2024
Log in to see stock. |
|||||
11831050
|
Hirschmann™
9000102 |
2 μL |
106.00 €
Pack of 1000 |
In Stock
Log in to see stock. |
|||||
10666751
|
Hirschmann™
9000130 |
30 μL |
113.00 €
Pack of 1000 |
Estimated Shipment: 11-07-2024
Log in to see stock. |
|||||
11841050
|
Hirschmann™
9000103 |
3 μL |
106.00 €
Pack of 1000 |
Estimated Shipment: 19-06-2024
Log in to see stock. |
|||||
11851050
|
Hirschmann™
9000104 |
4 μL |
106.00 €
Pack of 1000 |
Estimated Shipment: 29-05-2024
Log in to see stock. |
|||||
11757663
|
Hirschmann™
9000150 |
50 μL |
113.00 €
Pack of 1000 |
Estimated Shipment: 05-09-2024
Log in to see stock. |
|||||
11861050
|
Hirschmann™
9000105 |
5 μL |
105.00 €
Pack of 1000 |
Estimated Shipment: 29-05-2024
Log in to see stock. |
|||||
11732293
|
Hirschmann™
9000140 |
40 μL |
113.00 €
Pack of 1000 |
Estimated Shipment: 19-06-2024
Log in to see stock. |
|||||
10267980
|
Hirschmann™
9001133 |
3.33 μL |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Description
- Not heparinized
- End-to-end minicaps
- Acc. to ISO 9000, CE acc. IVD 98/79 EC
- Color-coded vial cap with capillary dispenser
- Conformity certified, with batch number
Spécification
Minicap Capillary Pipette | |
Single-use |