missing translation for 'onlineSavingsMsg'
Learn More

Macherey-Nagel™ Glass N11-1/N8-1 Small Opening Micro Insert

Product Code. 10658572 Shop All Macherey-Nagel Products
Change view
Click to view available options
Color:
Clear
Unit Size:
Pack of 100
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color Unit Size
10658572 Clear Pack of 100
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10658572 Supplier Macherey-Nagel™ Supplier No. 702968
ChemsChrom Mega Deals
Use promo code 27993
View all promotions
In Stock
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifikationer

Color Clear
Material Glass
Volume (Metric) 0.1 mL
Style For vials with Small opening
Base Style Conical
Quantity 100 Pk.
For Use With (Equipment) Small Opening Vials
Height (Metric) 31 mm
Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.