Learn More
Fisherbrand™ Glass Pear-Shaped Separating Funnel with Glass Stopcock
Separates mixtures into immiscible solvents. Fisherbrand™ Glass Pear-Shaped Separating Funnel with Glass Stopcock is available with interchangeable glass or PTFE stopcock, but supplied with a glass and plastic stopper.
Brand: Fisherbrand™
Code : 41
Additional Details : Weight : 0.17120kg
Description
Separates mixtures into immiscible solvents. Fisherbrand™ Glass Pear-Shaped Separating Funnel with Glass Stopcock is available with interchangeable glass or PTFE stopcock, but supplied with a glass and plastic stopper.
- Easy-open stopper helps to reduce pressure buildup during processing.
- Constructed in chemical resistant borosilicate glass.
- ISO 4800 Certified.
Specifications
50 mL | |
ISO 4800 | |
Glass | |
19/26 |
Borosilicate Glass | |
1PK | |
Glass, Plastic | |
Separating funnel |