Learn More
Hoefer™ Glass Plates for SE 600 Chroma, SE 600 Standard, or SE 400 Vertical Electrophoresis Units
Product Code. 11519753
Change view
Click to view available options
Type:
Glass Plate
Low Fluorescence Glass Plate
Unit Size:
Pack of 2
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Type Unit Size
11519753 Glass Plate Pack of 2
11905368 Low Fluorescence Glass Plate Pack of 2
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11519753 Supplier Hoefer™ Supplier No. SE6102
Estimated Shipment: 26-01-2027
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Accessory parts for SE 600 Chroma SE 600 Standard or SE 400 Vertical Electrophoresis Units
  • For use with SE660, SE600X Chroma, SE600, SE640, SE410, and SE400 Vertical Electrophoresis Units
  • Regular (straight, without notch)
  • Supplied in packs of 2
Specifications
For Use With (Equipment) SE 600 Chroma, SE 600 Standard, SE 400 Vertical Electrophoresis Unit
Type Glass Plate
Material Glass
Quantity 2/Pk.
Dimensions (L x W) 18 x 16 cm
Length (Metric) 18 cm
Width (Metric) 16 cm