Learn More
DCP Microdevelopments™ Glass Prepared Microscope Slides: Buttercup (Ranunculus) t.s. Mature Root
Product Code. 13153220
Encompass
Change view
Click to view available options
Quantity:
1 Ea.
Unit Size:
Each
Product Code. Quantity unitSize
13153220 1 Ea. Each
1 options
Product Code. 13153220 Supplier DCP Microdevelopments™ Supplier No. A11277
This item requires a price update prior to being ordered.
Request a Price for this item here.
Prepared slide of Ranunculus, (buttercup) t.s. mature root with metaxylem
Specifications
Material Glass
Type Pre-prepared Microscopic Slide
Quantity 1 Ea.
Species Ranunculus
Specimen Type Ranunculus mature root