Learn More
Poulten Selfe™ Glass Reverse Flow Viscometers
Product Code. 13141083
Change view
Click to view available options
Viscosity Range:
0.4 to 2 cSt
0.8 to 4 cSt
1.6 to 8 cSt
100 to 500 cSt
1600 to 8000 cSt
20 to 100 cSt
240 to 1200 cSt
3 to 15 cSt
4000 to 20,000 cSt
500 to 2500 cSt
7 to 35 cSt
Unit Size:
Each
11 product options available for selection
Product selection table with 11 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Viscosity Range unitSize
13141083 0.4 to 2 cSt Each
13151083 0.8 to 4 cSt -
13141093 1600 to 8000 cSt Each
13161083 1.6 to 8 cSt Each
13111093 100 to 500 cSt -
13191083 20 to 100 cSt Each
13121093 240 to 1200 cSt -
13171083 3 to 15 cSt -
13151093 4000 to 20,000 cSt Each
13131093 500 to 2500 cSt -
13181083 7 to 35 cSt -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 11 options available.
11 options
Product Code. 13141083 Supplier Poulten Selfe™ Supplier No. 1641/01
Suitable for measuring opaque liquids
Opaque 240 to 1,200v(mm2s-1,cst) 400 Poulten Selfe and Lee
Specifications
Product Type Reverse Flow Viscometer
Viscosity Range 0.4 to 2 cSt