Learn More
Poulten Selfe™ Glass Routine Viscometer
Product Code. 13181073
Change view
Click to view available options
Viscosity Range:
0.5 to 2 cSt
1600 to 8000 cSt
20 to 100 cSt
240 to 1200 cSt
500 to 2500 cSt
Unit Size:
Each
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Viscosity Range Unit Size
13181073 20 to 100 cSt Each
13141073 0.5 to 2 cSt Each
13131083 1600 to 8000 cSt Each
13111083 240 to 1200 cSt Each
13121083 500 to 2500 cSt Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Product Code. 13181073 Supplier Poulten Selfe™ Supplier No. 1634/06
  • ONLINE EXCLUSIVE PRICE
    183.00 €
    169.96 € / Pack of 1
    Save 13.04 €
    7% Off
 Restricted Item
Estimated Shipment: 17-11-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Suitable for measuring transparent liquids
ASTM D445-IP71, ASTM D446. Suitable for transparent liquids, nominal overall length 250mm. Approximate sample size 7mL. Calibration data at 40°C with the constant, C, stated at 40°C and 100°C. With NAMAS certification of calibration.
Specifications
Product Type Routine Viscometer
Viscosity Range 20 to 100 cSt
Calibrated Yes
Material Glass
Size 200