missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
- Excellent chemical resistance
- High temperature resistance
- Minimal thermal expansion giving relatively high resistance to temperature changes
- Cone Size: 24/29 NS
- Socket Size: 29/32 NS
- Borosilicate glass 3,3 clear
Specifications
Specifications
| Color | Clear |
| Cone Size | 24/29 NS |
| Description | Glass Sleeve Joints |
| For Use With (Application) | Expansion adapter from small cone to large socket. |
| Material | Borosilicate glass |
| Thread Size | Socket Size: 29/32 NS |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?