Learn More
Hirschmann™ Disposable Micro Capillary Pipettes, End-to-End
Facilitates error-free capillary filling in the laboratory. Hirschmann Instruments™ Disposable Micro Capillary Pipettes, End-to-End are manufactured from durable DURAN™ glass.
Brand:  Hirschmann™ 9000203
Code : A0
Additional Details : Weight : 0.80000kg
 View more versions of this product
Product Code. 11787663
  • 132.00 € / Pack of 1000
In Stock
to see stock.
Add to basket
Description
Description
  • End-to-end, with minicaps
  • Conformity certified
  • Available either heparinized or non-heparinized
  • Acc. to ISO 7550, CE acc. IVD 98/79 EC
Specifications
Specifications
0.5%
Disposable micro capillary pipettes, not heparinized, end-to-end; includes Pipette bulb (per 1000 pcs.)
10 x 100/Pack
Minicap Capillary Pipette
ISO 7550, CE acc. IVD 98/79 EG, conformity certified
Borosilicate Glass
3 μL
Single-use