Learn More
Glass Syringes
Product Code. 12309229
Click to view available options
Volume (Metric):
2 mL
10 mL
20 mL
Unit Size:
Each
Product Code. 12309229
Brand: S Murray SAMCO G812/05
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Use these syringes for a variety of applications. Glass syringes feature a metal tip and are fully interchangeable.
  • Glass syringe with metal tip
  • Luer fitting
  • Fully interchangeable
  • Available in a variety of sizes
Specifications
Material Glass
Product Type Syringe
Volume (Metric) 20 mL