Learn More
Poulten Selfe™ Glass Ubbelohde
Product Code. 13191063
Change view
Click to view available options
Size:
1B
2B
3
3C
4
Unit Size:
Each
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size Unit Size
13191063 2B Each
13161063 1B Each
13101073 3 Each
13111073 3C Each
13131073 4 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Product Code. 13191063 Supplier Poulten Selfe™ Supplier No. 1643/09
  • ONLINE EXCLUSIVE PRICE
    251.00 €
    215.02 € / Pack of 1
    Save 35.98 €
    14% Off
View all promotions
 Restricted Item
Estimated Shipment: 08-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Suitable for transparent liquids
ASTM D445-IP71, ASTM D446. Suitable for transparent liquids. Nominal overall length 283mm. Approximate sample size 18mL. With UKAS certificate of calibration.
Specifications
Product Type Ubbelohde Viscometer
Viscosity Range 100 to 500 cSt
Calibrated Yes
Material Glass
Size 2B