missing translation for 'onlineSavingsMsg'
Learn More

Foxx Life Sciences Glass Vacuum Trap System

Product Code. 31933504
Encompass_Preferred
Change view
Click to view available options
Capacity (Metric):
1 L (Primary) , 1 L (Overflow)
10 L (Primary), 2 L (Overflow)
2 L
2 L (Primary), 1 L (Overflow)
2 L (Primary), 2 L (Overflow)
3 L (Primary), 1 L (Overflow)
5 L
5 L (Primary), 2 L (Overflow)
Unit Size:
1 set
Each
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Unit Size
31933504 2 L (Primary), 2 L (Overflow) 1 set
31933395 1 L (Primary) , 1 L (Overflow) 1 set
31933351 2 L Each
31933454 2 L (Primary), 1 L (Overflow) 1 set
31933465 3 L (Primary), 1 L (Overflow) 1 set
31933385 5 L Each
31933421 5 L (Primary), 2 L (Overflow) 1 set
31933410 10 L (Primary), 2 L (Overflow) 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
This item is not returnable. View return policy
Product Code. 31933504 Supplier Foxx Life Sciences Supplier No. 30R2201FLS
 Restricted Item
Estimated Shipment: 19-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Designed to meet strict CDC laboratory guidelines for handling biohazardous waste

  • Features:
    • Vacuum trap assembly with chemical resistant glass bottles
    • 40 CFR 264.175 compliant aspirator system has a compact, tip-resistant secondary container for stability
    • Bottles made with BOROSILICATE 3.3 are chemically inert and ideal for harsh chemicals
    • Standard vacuum trap with GL45 VersaCap™ system with rotating adapter prevents tube tangling
    • Primary bottle volume: 1 L, Overflow bottle volume: 1 L
    • Made in USA
  • Applications:
    • Aspiration of cell culture media
    • Vacuum pump trap kit
    • Waste collection system for high-purity processes
    • Aspirator bottle assembly
    • Ideal for house vacuum or vacuum pump trap systems

Specifications

Capacity (Metric) 2 L (Primary), 2 L (Overflow)
Case Material Polypropylene
Material Borosilicate Glass
Maximum Pressure 1 bar
Includes Secondary Container, Bottles, Cap Assembly with Vent Filter, Tubing, Pinch Clamp
Type Glass Vacuum Trap System
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.