missing translation for 'onlineSavingsMsg'
Learn More

Hecht Karl™ Glass Wide Mouth Reagent Bottle

Product Code. 10093751
Change view
Click to view available options
Capacity (Metric):
100 mL
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Unit Size
10093751 100 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10093751 Supplier Hecht Karl™ Supplier No. 41311012
  • ONLINE EXCLUSIVE PRICE
    10.50 €
    7.22 € / Pack of 1
    Save 3.28 €
    31% Off
 Restricted Item
Estimated Shipment: 27-01-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Note: Glass plug has grooves, as shown in images 2 and 3 above.

Specifications

Capacity (Metric) 100 mL
Color Clear
Material Glass
Closure Type Glass plug
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.