Learn More
Heidolph™ Glassware Set - Diagonal Condenser (Standard Distillations)
Shop All Heidolph Instruments ProductsBeschreibung
- Glassware sets with a transparent plastic coating are suitable for a temperature range up to max. 80™C
- Glassware set includes (1) 1000 mL evaporating flask and (1) 1000 mL receiving flask
- Evaporating flasks and vapor tubes come with a standard NS 29/32 joint size but are also available with NS 24/29
- Ventilation cap without ground joints for easy handling and grease-free operation included
- A venting valve with refill function is optionally available
- Not suitable for the automated distillation program Dynamic AUTOaccurate on Hei-VAP Expert/Ultimate Control models
- Also available upon request: glassware sets for Soxhlet extraction and descending condenser system
Spezifikation
Spezifikation
| Material | Glass |
| Type | Glassware Set |
| For Use With (Equipment) | Rotary Evaporators |
| Includes | Glassware set G1 - Diagonal condenser (all standard distillations), 1000 mL evaporating flask and 1000 mL receiving flask |
| Material (Coating) | Uncoated |
| Volume (Metric) | 1000 mL |