Learn More
Heidolph™ Glassware Set - Dry Ice Condenser (Low-boiling Solvents)
Product Code. 10690303 Shop All Heidolph Instruments Products
Change view
Click to view available options
Volume (Metric):
1000 mL
Unit Size:
Each
Product Code. Volume (Metric) Unit Size
10690303 1000 mL Each
1 options
Product Code. 10690303 Supplier Heidolph™ Supplier No. 5130050000
Equipment and Instrument End of Year Sale
Use promo code 28000
View all promotions
Estimated Shipment: 24-11-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Dry Ice condenser for low-boiling solvents
  • Glassware set includes (1) 1000 mL evaporating flask and (1) 1000 mL receiving flask
  • Evaporating flasks and vapor tubes come with a standard NS 29/32 joint size but are also available with NS 24/29
  • Ventilation cap without ground joints for easy handling and grease-free operation included
  • A venting valve with refill function is optionally available
  • Not suitable for the automated distillation program Dynamic AUTOaccurate on Hei-VAP Expert/Ultimate Control models
  • Also available upon request: glassware sets for Soxhlet extraction and descending condenser system
TRUSTED_SUSTAINABILITY
Specifications
Material Glass
Type Glassware Set
For Use With (Equipment) Rotary Evaporators
Includes Glassware set G5 - Dry ice condenser (low boiling solvents), 1000 mL evaporating flask and 1000 mL receiving flask
Material (Coating) Uncoated
Volume (Metric) 1000 mL