Learn More
Heidolph™ Glassware Set - Vertical Condenser (Standard Distillation)
Product Code. 10219271 Shop All Heidolph Instruments Products
Change view
Click to view available options
Volume (Metric):
1000 mL
Unit Size:
Each
Product Code. Volume (Metric) Unit Size
10219271 1000 mL Each
1 options
Product Code. 10219271 Supplier Heidolph™ Supplier No. 5130031000
Estimated Shipment: 17-11-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Vertical condenser for all standard distillations is the option with the smallest space requirement
  • Glassware sets with a transparent plastic coating are suitable for a temperature range up to max. 80™C
  • Glassware set includes (1) 1000 mL evaporating flask and (1) 1000 mL receiving flask
  • Evaporating flasks and vapor tubes come with a standard NS 29/32 joint size but are also available with NS 24/29
  • Ventilation cap without ground joints for easy handling and grease-free operation included
  • A venting valve with refill function is optionally available
  • Not suitable for the automated distillation program Dynamic AUTOaccurate on Hei-VAP Expert/Ultimate Control models
  • Also available upon request: glassware sets for Soxhlet extraction and descending condenser system
TRUSTED_SUSTAINABILITY
Specifications
Material Glass
Type Glassware Set
For Use With (Equipment) Rotary Evaporators
Includes Glassware set G3B - Vertical condenser (all standard distillations), 1000 mL evaporating flask and 1000 mL receiving flask with transparent plastic coating
Material (Coating) Transparent plastic
Volume (Metric) 1000 mL