Learn More
GLI-2 Sheep anti-Human, Polyclonal, R&D Systems™
GREENER_CHOICE
Sheep Polyclonal Antibody has been used in 3 publications
Brand:  R&D Systems AF3526-SP
Encompass
Product Code. 15752074
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Description
Description
GLI-2 Polyclonal specifically detects GLI-2 in Human samples. It is validated for Western Blot, Immunohistochemistry, Chromatin Immunoprecipitation (ChIP).