missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Goat anti-Mouse IgG F(ab) Secondary Antibody, FITC, Novus Biologicals™
Goat Polyclonal Antibody
Brand: Novus Biologicals NBP2-68470F
This item is not returnable.
View return policy
Description
IgG F(ab) Polyclonal antibody specifically detects IgG F(ab) in Mouse samples. It is validated for Western Blot, ELISA
Specifications
| IgG F(ab) | |
| Polyclonal | |
| PBS | |
| Mouse IgG Fab fragments | |
| 1 mL | |
| Secondary | |
| Store at 4°C in the dark. | |
| IgG |
| Western Blot, ELISA | |
| FITC | |
| Goat | |
| Affinity purified | |
| RUO | |
| Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction