missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Brady™ GlobalMark™ Single Color Labeling Ribbons
Brand: Brady™ 800523
Code : 50
Additional Details : Weight : 0.38600kg
Specifications
105 mm Red Ribbon | |
Single Color Labeling Ribbon | |
GlobalMark™ printer | |
60 m x 105 mm | |
105 mm |
Red | |
Printed text last for years outdoors and in industrial areas | |
1 roll/pk | |
60 m |