Learn More
Globe Scientific Polypropylene Sample Cup for Alpha Wassermann™ ACE and Gilford™ Analyzers
Product Code. 31922712 Shop All Globe Scientific Products
Change view
Click to view available options
For Use With (Equipment):
Alpha Wassermann ACE and Gilford Analyzers
Unit Size:
Pack of 1000
Product Code. For Use With (Equipment) Unit Size
31922712 Alpha Wassermann ACE and Gilford Analyzers Pack of 1000
1 options
Product Code. 31922712 Supplier Globe Scientific Supplier No. 5545
Estimated Shipment: 29-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
For use with Alpha Wassermann ACE and Gilford analyzers.
  • Capacity: 0.5mL
  • Dimensions: 0.3 x 0.8 in. (10 x 22mm)
Specifications
Type Sample Cup
For Use With (Equipment) Alpha Wassermann ACE and Gilford Analyzers
Capacity (Metric) 5 mL
Quantity 1000/Pk.