Learn More
Globe Scientific Accessories for Sysmex™ Analyzers
Product Code. 31922793 Shop All Globe Scientific Products
Change view
Click to view available options
For Use With (Equipment):
Sysmex CA 50, 500, 1000, 1500, and 5000 analyzers
Unit Size:
Pack of 1000
Product Code. For Use With (Equipment) Unit Size
31922793 Sysmex CA 50, 500, 1000, 1500, and 5000 analyzers Pack of 1000
1 options
Product Code. 31922793 Supplier Globe Scientific Supplier No. 5531
Estimated Shipment: 29-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
For use with Sysmex analyzers.
  • For use with Sysmex CA 50, 500, 1000, 1500, and 5000 analyzers
  • Material: Polystyrene
  • Capacity: 2mL
Specifications
Type Sample Cup
For Use With (Equipment) Sysmex CA 50, 500, 1000, 1500, and 5000 analyzers
Capacity (Metric) 2 mL
Quantity 1000/Pk.