Learn More
Globe Scientific Coagulation Cup for Accu Coa Data Coagulation Analyzers
Product Code. 31923537 Shop All Globe Scientific Products
Change view
Click to view available options
For Use With (Equipment):
Accu Coa Data Coagulation Analyzers
Unit Size:
Pack of 5000
Product Code. For Use With (Equipment) Unit Size
31923537 Accu Coa Data Coagulation Analyzers Pack of 5000
1 options
Product Code. 31923537 Supplier Globe Scientific Supplier No. 5574
Estimated Shipment: 29-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
For use on coagulation instruments.
  • Made of Polystyrene
  • Supplied with metal mixing bars
  • Dimensions: 1.3 x 0.3 in. (35.1 x 8.6mm)
Specifications
Type Coagulation Cup
For Use With (Equipment) Accu Coa Data Coagulation Analyzers
Includes Metal mixing bars