Learn More
Globe Scientific Pediatric Micro Sample Cup
Product Code. 31923105
Change view
Click to view available options
Material:
Polystyrene
Unit Size:
Pack of 6000
Product Code. Material Unit Size
31923105 Polystyrene Pack of 6000
1 options
Product Code. 31923105 Supplier Globe Scientific Supplier No. 110915
Estimated Shipment: 30-11-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
For use on automated instrumentation for Roche™ analyzers
  • Made of polystyrene
  • Also for use on Hitachi, Elecsys, Cobas Integra and Core analyzers
  • Dimensions: 0.6 x 1.45 in. (17 x 37mm)
Specifications
Product Type Sample Cup
Material Polystyrene
For Use With (Equipment) Automated Roche™, Hitachi™, Elecsys™, Cobas Integra™ and Core™ analyzers