Learn More
Thermo Scientific Chemicals Glove Bag Clamp
Product Code. 15491837
Change view
Click to view available options
Quantity:
1 Ea.
Unit Size:
Each
Product Code. Quantity Unit Size
15491837 1 Ea. Each
1 options
Product Code. 15491837 Supplier Thermo Scientific Chemicals Supplier No. 042909.KF
 Restricted Item
Estimated Shipment: 26-11-2026
Add to basket
missing translation for 'validMainframeMsg'
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
  • Quick-closing clamp
  • For use with 17 x 17 in. polyethylene glove bags
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) Polyethylene Glove Bags (17 x 17 in.)
Quantity 1 Ea.