missing translation for 'onlineSavingsMsg'
Learn More

Labconco™ Glove Box Gloves, Sleeves and Hands

Product Code. 17951217 Shop All Labconco Corporation Products
Change view
Click to view available options
Material:
Butyl
CSM
Neoprene
Size:
8.5
9.75
Thickness (English):
15 mil
Color:
Black
White
Unit Size:
1 pair
Each
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Material Size Thickness (English) Color Unit Size
17951217 CSM 9.75 15 mil White 1 pair
16467130 Butyl 9.75 15 mil Black Each
17941217 CSM 8.5 15 mil White 1 pair
11781489 Neoprene 9.75 15 mil Black Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 17951217 Supplier Labconco™ Supplier No. 5005602
  • ONLINE EXCLUSIVE PRICE
    659.00 €
    622.75 € / 1 pairs
    Save 36.25 €
    5% Off
 Restricted Item
Estimated Shipment: 19-01-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Glove box gloves, sleeves and hands are available in a variety of sizes, materials and thicknesses to suit your specific application. Choose from neoprene, latex, CSM or butyl.

Glove Box Gloves - One pair/pack, one-piece, seamless, with O-rings

  • Neoprene - Size 9.75
    • 17 mil thickness
  • CSM (Chlorosulfonated polyethylene rubber) - Available in sizes 8.5 or 9.75
    • Also known as Hypalon
    • Resistant to abrasion, ozone and oxidizing chemicals
    • 15 mil thickness
  • Butyl - Available in sizes 8.5 and 9.75
    • Provides high impermeability to water vapor, gases and toxic chemicals
    • Resistant to oxygenated solvents and most oxidizing chemicals
    • 15 mil thickness

Glove Box Sleeves

  • Neoprene Straight Sleeves
    • For use with Neoprene or Latex Hands
    • With O-rings and springs

Glove Box Hands - For use with neoprene straight or accordian sleeves

  • Neoprene available in size 7.5, 9 or 10
    • 17 mil thickness
    • Lightweight and embossed with firm hold finish
  • Latex available in size 7 or 9
    • 9 mil thickness
    • White, surgical-type

Specifications

Color White
Hand Ambidextrous
Latex Free Yes
Material CSM
Product Type Glove Box Gloves
Size 9.75
Sterility Non-sterile
Thickness (English) 15 mil
Manufacturing/QMS Audit Standards ISO 9001
Model 5005602
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.