missing translation for 'onlineSavingsMsg'
Learn More

GLPG-0492, TRC

Product Code. 30006775
Change view
Click to view available options
Quantity:
1 mg
2.5 mg
5 mg
Unit Size:
1mg
2.5mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30006775 5 mg 5mg
30004473 2.5 mg 2.5mg
30006426 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30006775 Supplier Toronto Research Chemicals Supplier No. TRCG4110005MG
 Restricted Item
Estimated Shipment: 01-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1215085-92-9
Molecular Formula C19H14F3N3O3
Molecular Weight (g/mol) 389.33
Synonym 4-[(4S)-4-(Hydroxymethyl)-3-methyl-2,5-dioxo-4-phenyl-1-imidazolidinyl]-2-(trifluoromethyl)benzonitrile
IUPAC Name 4-[(4S)-4-(hydroxymethyl)-3-methyl-2,5-dioxo-4-phenylimidazolidin-1-yl]-2-(trifluoromethyl)benzonitrile
SMILES CN1C(=O)N(C2=CC(=C(C=C2)C#N)C(F)(F)F)C(=O)[C@@]1(CO)C3=CC=CC=C3

Specifications

Quantity 5 mg
InChI Formula InChI=1S/C19H14F3N3O3/c1-24-17(28)25(14-8-7-12(10-23)15(9-14)19(20,21)22)16(27)18(24,11-26)13-5-3-2-4-6-13/h2-9,26H,11H2,1H3/t18-/m1/s1
Formula Weight 389.0987
Chemical Name or Material GLPG-0492
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.