missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Glucagon-Like Peptide-1 (7-36) amide, human

Product Code. 15477459
Click to view available options
Quantity:
0.5 mg
1 mg
Unit Size:
0.5mg
1mg
This item is not returnable. View return policy

Product Code. 15477459

Brand: Thermo Scientific Chemicals J66197.!D

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

For cellular and molecular biology applications

Glucagon-Like Peptide-1 is used as a Effector in the hormonal control of insulin secretion. The peptide fragment is secreted from the lower small intestine. Its action is mediated by receptors expressed by the endocrine pancreatic B-cells.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

Species Human
Purification Method HPLC
Common Name Preproglucagon (78-107) amide, human
Content And Storage -30°C to -10°C
Format Powder
Molecular Weight (g/mol) 3297.70
Product Type Glucagon-Like Peptide-1 (7-36) amide, human
Quantity 0.5 mg
Regulatory Status RUO - research use only
Sequence GLP-1 (7-36) amide, human

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.