missing translation for 'onlineSavingsMsg'
Learn More

Glucametacin, TRC

Product Code. 30407225
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30407225 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30407225 Supplier Toronto Research Chemicals Supplier No. TRCG41280010MG
 Restricted Item
Estimated Shipment: 14-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Glucametacin
CAS 871014-84-5
Molecular Formula C25H27ClN2O8
Quantity 10 mg
Synonym 2-[[[1-(4-Chlorobenzoyl)-5-methoxy-2-methyl-1H-indol-3-yl]acetyl]amino]-2-deoxy-α-D-Glucopyranose, α-Glucametacine, α-Glucamethacin, α-Indomethacin Glucosamide
InChI Formula InChI=1S/C25H27ClN2O8/c1-12-16(10-20(30)27-21-23(32)22(31)19(11-29)36-25(21)34)17-9-15(35-2)7-8-18(17)28(12)24(33)13-3-5-14(26)6-4-13/h3-9,19,21-23,25,29,31-32,34H,10-11H2,1-2H3,(H,27,30)/t19-,21-,22-,23-,25+/m1/s1
SMILES CC1=C(C2=C(N1C(=O)C3=CC=C(C=C3)Cl)C=CC(=C2)OC)CC(=O)N[C@@H]4[C@H]([C@@H]([C@H](O[C@@H]4O)CO)O)O
IUPAC Name 2-[1-(4-chlorobenzoyl)-5-methoxy-2-methylindol-3-yl]-N-[(2S,3R,4R,5S,6R)-2,4,5-trihydroxy-6-(hydroxymethyl)oxan-3-yl]acetamide
Molecular Weight (g/mol) 518.94
Formula Weight 518.1456
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.