missing translation for 'onlineSavingsMsg'
Learn More

Gluconate Calcium, MedChemExpress

Product Code. 16428606
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16428606 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16428606 Supplier Medchem Express Supplier No. HYB1092
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

MedChemExpress Gluconate Calcium (Calcium D-gluconate) is a mineral supplement, manufactured by the neutralization of gluconic acid with lime or calcium carbonate.

Gluconate Calcium (Calcium D-gluconate) is a mineral supplement, manufactured by the neutralization of gluconic acid with lime or calcium carbonate.

Specifications

Chemical Name or Material Gluconate Calcium
CAS 299-28-5
Color White
Purity Grade Notes Research
Molecular Formula C6H12O7.1/2Ca
Quantity 100 mg
Synonym Calcium D-gluconate Gluconic acid hemicalcium salt
Solubility Information H2O : 20 mg/mL (92.51 mM; Need ultrasonic) ∣DMSO : 1 mg/mL (4.63 mM; ultrasonic and warming and heat to 80°C)
SMILES O[C@H]([C@H]([C@@H]([C@@H](CO)O)O)O)C(O)=O.[1/2Ca]
Molecular Weight (g/mol) 216.2
Formula Weight 216.2
Percent Purity 98.0%
Grade Research
Physical Form Solid
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.