missing translation for 'onlineSavingsMsg'
Learn More

Gluconic acid, sodium salt, 98%

Product Code. 10175773 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
1 kg
2.5 kg
10 kg
Packaging:
Glass Bottle
Plastic Bottle
Plastic drum
Unit Size:
10kg
1kg
2.5kg
5g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10175773 10 kg Plastic drum 10kg
10733571 5 g Glass Bottle 5g
10582011 1 kg Plastic Bottle 1kg
10604692 2.5 kg Plastic Bottle 2.5kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 10175773 Supplier Thermo Scientific Acros Supplier No. 181390100
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 527-07-1 | C6H11NaO7 | 218.137 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 527-07-1
Molecular Formula C6H11NaO7
Molecular Weight (g/mol) 218.137
MDL Number MFCD00064210
InChI Key UPMFZISCCZSDND-JJKGCWMISA-M
Synonym sodium gluconate, sodium d-gluconate, d-gluconic acid sodium salt, d-gluconic acid, monosodium salt, gluconic acid sodium salt, monosodium gluconate, glonsen, monosodium d-gluconate, gluconate sodium, pasexon 100t
PubChem CID 23672301
ChEBI CHEBI:84997
IUPAC Name sodium;(2R,3S,4R,5R)-2,3,4,5,6-pentahydroxyhexanoate
SMILES C(C(C(C(C(C(=O)[O-])O)O)O)O)O.[Na+]

Specifications

Melting Point 206.0°C to 209.0°C
Color White to Beige
pH 6.5 to 7.5 (10% soln.)
Infrared Spectrum Authentic
Assay Percent Range 0.98
Packaging Plastic drum
Linear Formula HOCH2[CH(OH)]4CO2Na
Quantity 10 kg
Beilstein 03, I, 188
Merck Index 15, 4492
Specific Rotation Condition + 12.10 (23.00°C c=1.75,H2O)
Specific Rotation + 12.10
Formula Weight 218.14
Percent Purity 0.98
Physical Form Crystalline Powder
Chemical Name or Material Gluconic acid, sodium salt
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.