missing translation for 'onlineSavingsMsg'
Learn More

Glucose-6-phosphate dehydrogenase

Product Code. 15443809
Change view
Click to view available options
Quantity:
1 kU
Packaging:
Plastic bottle
Unit Size:
1kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
15443809 1 kU Plastic bottle 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15443809 Supplier Thermo Scientific Alfa Aesar Supplier No. J61181.4I
Curiosity Goes Beyond
Use promo code 27966
View all promotions
 Restricted Item
In Stock
Add to basket
Edge
This item is not returnable. View return policy

CAS: 9001-40-5

Glucose-6-phosphate dehydrogenase, yeast converts p-glucose-6-phosphate to p-glucono-d-lactone-6-phosphate in presence of NADP+ or NAD+ for the determination of blood glucose or creatine kinase. It is used to investigate the effect of dehydroepiandrosterone on cell growth and mitochondrial function in TM-3 cells, in bovine follicular cells and in the incidence of apoptosis and transcript abundance. It is also useful to test ketose reductase activity in developing maize endosperm.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 9001-40-5
Packaging Plastic bottle
MDL Number MFCD00081656
Quantity 1 kU
Synonym EC 1.1.1.49,RP-87086
Solubility Information Soluble in water and dilute buffer.
Physical Form Lyophilized Powder
Chemical Name or Material Glucose-6-phosphate dehydrogenase

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.