missing translation for 'onlineSavingsMsg'
Learn More

Glucose monohydrate, Solstice
BIOPHARMA

Product Code. 16666153 Shop All Honeywell Fluka Products
Change view
Click to view available options
Quantity:
1 kg
2.5 kg
6 x 1 kg
6 x 2.5 kg
Unit Size:
1kg
2.50kg
Pack of 6
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16666153 6 x 2.5 kg Pack of 6
16636153 1 kg 1kg
16646153 2.5 kg 2.50kg
16656153 6 x 1 kg Pack of 6
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16666153 Supplier Fluka Supplier No. 16301H6X2.5KG
  • ONLINE EXCLUSIVE PRICE
    470.00 €
    441.97 € / Pack of 6
    Save 28.03 €
    5% Off
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

Currently Unavailable: We have no information when this product will be available.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Meets analytical specification of Ph. Eur., BP, Ph Franç., 7.0-9.5% water

  • Wide selection of support materials and multiple cartridge/column sizes
  • Every stationary phase batch tested to ensure lot-to-lot reproducibility
  • B&J’s reliable quality

 

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10-1-5996
Molecular Formula C6H14O7
Molecular Weight (g/mol) 198.17
InChI Key OSNSWKAZFASRNG-WNFIKIDCSA-N
PubChem CID 133126654
IUPAC Name (2S,3R,4S,5S,6R)-6-(hydroxymethyl)oxane-2,3,4,5-tetrol hydrate
SMILES O.OC[C@H]1O[C@H](O)[C@H](O)[C@@H](O)[C@@H]1O

Specifications

Packaging Plastic Bottle
Quantity 6 x 2.5 kg
Residual Solvents Complying
Percent Purity 97.5 to 102.0%
Grade Reagent Pharmacopeia Organics
Physical Form Powder
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.