missing translation for 'onlineSavingsMsg'
Learn More

Glutacondianil Hydrochloride, TRC

Product Code. 30399138
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30399138 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30399138 Supplier Toronto Research Chemicals Supplier No. TRCG5969305G
 Restricted Item
Estimated Shipment: 26-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Glutacondianil Hydrochloride
CAS 1497-49-0
Molecular Formula C17H16N2 . HCl
Quantity 5 g
Synonym N-[5-(Phenylamino)-2,4-pentadien-1-ylidene]benzenamine Hydrochloride, N,N'-1,3-Pentadien-1-yl-5-ylidenedianiline Hydrochloride, N,N'-1,3-Pentadien-1-yl-5-ylidenedianiline Monohydrochloride, N-[5-(Phenylamino)-2,4-pentadienylidene]benzenamine Monohydrochloride, 1-Phenylamino-5-phenylimino-1,3-pentadiene Hydrochloride, Glutaconaldehyde Dianil Chloride, Glutaconaldehyde Dianilide Hydrochloride, Glutaconic Aldehyde Dianil Hydrochloride, Pentadienedianiline Hydrochloride
InChI Formula InChI=1S/C17H16N2.ClH/c1-4-10-16(11-5-1)18-14-8-3-9-15-19-17-12-6-2-7-13-17;/h1-15,18H;1H/b9-3+,14-8+,19-15+;
SMILES Cl.N(\C=C\C=C\C=N\c1ccccc1)c2ccccc2
IUPAC Name N-[(1E,3E,5E)-5-phenyliminopenta-1,3-dienyl]aniline;hydrochloride
Molecular Weight (g/mol) 284.8
Formula Weight 284.108
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.