missing translation for 'onlineSavingsMsg'
Learn More

Glutaconic aldehyde dianilide hydrochloride, mixture of isomers, 98%

Product Code. 11488317
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11488317 100 g 100g
11468317 5 g 5g
11478317 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11488317 Supplier Thermo Scientific Alfa Aesar Supplier No. A18390.22
 Restricted Item
Estimated Shipment: 11-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 1497-49-0 | C17H17ClN2 | 284.79 g/mol

Glutaconic aldehyde dianilide hydrochloride is a tricarbocyanine laser dye intermediate. It is used as an intermediate for pharmaceuticals and dyes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Glutaconic aldehyde dianilide hydrochloride is a tricarbocyanine laser dye intermediate. It is used as an intermediate for pharmaceuticals and dyes.

Solubility
Soluble in methanol. Insoluble in water..

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents. It is hygroscopic in nature.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1497-49-0
Molecular Formula C17H17ClN2
Molecular Weight (g/mol) 284.79
MDL Number MFCD00012630
InChI Key VUCMMJBDNXZQDJ-ZUJIUJENSA-N
Synonym hydrogen n-1e,3e-5-phenylimino penta-1,3-dien-1-yl aniline chloride
PubChem CID 5702667
IUPAC Name N-[(1E,3E)-5-phenyliminopenta-1,3-dienyl]aniline;hydrochloride
SMILES [H+].[Cl-].N(\C=C\C=C\C=NC1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point ∼124°C to 132°C
Quantity 100 g
Beilstein 3916924
Sensitivity Hygroscopic
Solubility Information Soluble in methanol. Insoluble in water..
Formula Weight 284.79
Percent Purity 98%
Chemical Name or Material Glutaconic aldehyde dianilide hydrochloride, mixture of isomers

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.