missing translation for 'onlineSavingsMsg'
Learn More

Glutaraldehyde, 50% aq. soln.

Product Code. 11438743
Change view
Click to view available options
Quantity:
100 g
500 g
2500 g
Unit Size:
100g
2500g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11438743 500 g 500g
11428743 100 g 100g
11448743 2500 g 2500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11438743 Supplier Thermo Scientific Alfa Aesar Supplier No. A10500.36
Follow the Workflow - Explore the Savings
Use promo code 27967
View all promotions
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 111-30-8 | C5H8O2 | 100.12 g/mol

Glutaraldehyde is used as a cross linking agent for gelatin, poly(vinyl alcohol) and polyheptapeptides. It is also used as a fixative for electron microscopy and as disinfectants. It provides water resistance to protein and polyhydroxy compounds by reacting through cross linking. It acts as an intermediate for the production of resins, dyestuffs, pharmaceuticals, photographic films and for stabilization of proteins on agarose beads, activation of polystyrene and glass for immobilization of antibodies and antigens and coupling peptides onto carrier proteins.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Glutaraldehyde is used as a cross linking agent for gelatin, poly(vinyl alcohol) and polyheptapeptides. It is also used as a fixative for electron microscopy and as disinfectants. It provides water resistance to protein and polyhydroxy compounds by reacting through cross linking. It acts as an intermediate for the production of resins, dyestuffs, pharmaceuticals, photographic films and for stabilization of proteins on agarose beads, activation of polystyrene and glass for immobilization of antibodies and antigens and coupling peptides onto carrier proteins.

Solubility
Miscible with water, acetone, dichloromethane, ethylacetate, isopropanol, n-hexane and toluene.

Notes
Incompatible with strong oxidisng agents, strong acids and strong bases.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 111-30-8
Molecular Formula C5H8O2
Molecular Weight (g/mol) 100.12
MDL Number MFCD00007025
InChI Key SXRSQZLOMIGNAQ-UHFFFAOYSA-N
Synonym glutaraldehyde, glutaral, glutaric dialdehyde, cidex, glutardialdehyde, 1,5-pentanedial, sonacide, glutarol, aldesan, glutaric aldehyde
PubChem CID 3485
ChEBI CHEBI:64276
IUPAC Name pentanedial
SMILES O=CCCCC=O

Specifications

Melting Point ∽-21°C
Density 1.123 g/mL
Boiling Point 100°C
Refractive Index 1.421
Quantity 500 g
Beilstein 605390
Solubility Information Miscible with water,acetone,dichloromethane,ethylacetate,isopropanol,n-hexane and toluene.
Formula Weight 100.12
Chemical Name or Material Glutaraldehyde
compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • Glutaraldehyde, 50% aqueous solution
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 2
  • Acute toxicity Category 3
  • LONG-TERM AQUATIC HAZARD Chronic 2
  • ACUTE AQUATIC Acute 1
  • Serious eye damage/eye irritation Category 1
  • Respiratory sensitiser Category 1
  • Skin corrosion/irritation Category 1 B
  • Skin sensitiser Category 1
  • Specific target organ toxicity Category 3
Hazard Statement
  • H301-Toxic if swallowed.
  • H314-Causes severe skin burns and eye damage.
  • H317-May cause an allergic skin reaction.
  • H318-Causes serious eye damage.
  • H330-Fatal if inhaled.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
  • H335-May cause respiratory irritation.
  • H400-Very toxic to aquatic life.
  • H411-Toxic to aquatic life with long lasting effects.
  • EUH071-Corrosive to the respiratory tract.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contain: Glutaraldehyde

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.